4-Chlorbenzophenon
Lieferant: Molekula
Synonyme:
p-Chlorobenzophenone
M45903721
MOLEM45903721EA
0
EUR
MOLEM45903721
4-Chlorbenzophenon
4-Chlorbenzophenon
Formel:
C₁₃H₉ClO Molecular Weight: 216,67 g/mol Siedepunkt: 333 °C (1013 hPa) Schmelzpunkt: 68…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000622 CAS-Nummer: 134-85-0 EINECS: 205-160-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...