(S)-(+)-1-Phenyl-1,2-ethandiol
Lieferant: Molekula
Synonyme:
(S)-(+)-α,β-Dihydroxyethylbenzol, (S)-(+)-(1,2-Dihydroxyethyl)benzol, (S)-(+)-Phenylethylenglycol, (S)-(+)-Phenylethan-1,2-diol
M46008201
MOLEM46008201EA
0
EUR
MOLEM46008201
(S)-(+)-1-Phenyl-1,2-ethandiol
(S)-(+)-1-Phenyl-1,2-ethandiol
Formel:
C₈H₁₀O₂ Molecular Weight: 138,17 g/mol Siedepunkt: 272 °C (1013 hPa) Schmelzpunkt: 67 °C |
MDL:
MFCD00066256 CAS-Nummer: 25779-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...