5-Chlor-2-nitroanisol

Lieferant: Molekula

M46260108
MOLEM46260108EA 0 EUR
MOLEM46260108
5-Chlor-2-nitroanisol
5-Chlor-2-nitroanisol
Formel: C₇H₆ClNO₃
Molecular Weight: 187,58 g/mol
Schmelzpunkt: 71 °C
MDL: MFCD00007288
CAS-Nummer: 6627-53-8
EINECS: 229-594-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR