5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na)
Lieferant: Molekula
Synonyme:
5-Bromo-4-chloro-1H-indol-3-yl phosphate sodium, X-phosphate disodium salt, Sodium 5-bromo-4-chloro-1H-indol-3-yl phosphate
M46642783
MOLEM46642783EA
0
EUR
MOLEM46642783
5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na)
5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na)
Formel:
C₈H₄BrClNNa₂O₄P Molecular Weight: 370.43 g/mol |
MDL:
MFCD00036757 CAS-Nummer: 102185-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...