Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)
Lieferant: Molekula
Synonyme:
Fmoc-OSu, N-(9-Fluorenylmethoxycarbonyloxy)succinimide
M46709264
MOLEM46709264EA
0
EUR
MOLEM46709264
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)
Formel:
C₁₉H₁₅NO₅ Molecular Weight: 337,3 g/mol |
MDL:
MFCD00010733 CAS-Nummer: 82911-69-1 EINECS: 433-520-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...