Ethyl-4-chloracetoacetat

Lieferant: Molekula

Synonyme: Ethyl 4-chloro-3-oxobutanoate

M46740896
MOLEM46740896EA 0 EUR
MOLEM46740896
Ethyl-4-chloracetoacetat
Ethyl-4-chloracetoacetat
Formel: C₆H₉ClO₃
Molecular Weight: 164,59 g/mol
MDL: MFCD00000939
CAS-Nummer: 638-07-3
EINECS: 211-317-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR