Theophyllin
Lieferant: Molekula
Synonyme:
1,3-Dimethyl-1H-purine-2,6(3H,9H)-dione, Theophylline
M47373244
MOLEM47373244EA
0
EUR
MOLEM47373244
Theophyllin
Theophyllin
Formel:
C₇H₈N₄O₂ Molecular Weight: 180,16 g/mol |
MDL:
MFCD00079619 CAS-Nummer: 58-55-9 EINECS: 200-385-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...