2-Benzo[b]thiophenboronsäure
Lieferant: Molekula
Synonyme:
Benzo[b]thiophen-2-ylboronic acid, 2-Benzothienylboronic acid
M47873272
MOLEM47873272EA
0
EUR
MOLEM47873272
2-Benzo[b]thiophenboronsäure
2-Benzo[b]thiophenboronsäure
Formel:
C₈H₇BO₂S Molecular Weight: 178,02 g/mol |
MDL:
MFCD01075674 CAS-Nummer: 98437-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...