2,5-Dimethoxybenzylalkohol

Lieferant: Molekula

M47897739
MOLEM47897739EA 0 EUR
MOLEM47897739
2,5-Dimethoxybenzylalkohol
2,5-Dimethoxybenzylalkohol
Formel: C₉H₁₂O₃
Molecular Weight: 168,19 g/mol
Siedepunkt: 122 °C (1013 hPa)
MDL: MFCD00004615
CAS-Nummer: 33524-31-1
EINECS: 251-562-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR