(3,4-Dimethoxyphenyl)acetonitril
Lieferant: Molekula
Synonyme:
Homoveratronitril, 3,4-Dimethoxybenzylcyanid
M47923583
MOLEM47923583EA
0
EUR
MOLEM47923583
(3,4-Dimethoxyphenyl)acetonitril
(3,4-Dimethoxyphenyl)acetonitril
Formel:
C₁₀H₁₁NO₂ Molecular Weight: 177.2 Siedepunkt: 320 Schmelzpunkt: 60…62 °C |
MDL:
MFCD00001911 CAS-Nummer: 93-17-4 EINECS: 202-225-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...