4-Bromresorcin
Lieferant: Molekula
Synonyme:
4-Bromresorcinol
M48786774
MOLEM48786774EA
0
EUR
MOLEM48786774
4-Bromresorcin
4-Bromresorcin
Formel:
C₆H₅BrO₂ Molecular Weight: 189,01 g/mol Siedepunkt: 273 °C (1013 hPa) Schmelzpunkt: 98 °C |
MDL:
MFCD00002272 CAS-Nummer: 6626-15-9 EINECS: 229-586-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...