Bis(4-hydroxyphenyl)sulfon

Lieferant: Molekula

Synonyme: 4,4'-Dihydroxydiphenylsulfon, 4-Hydroxyphenylsulfon, 4,4'-Sulfonyldiphenol

M48860052
MOLEM48860052EA 0 EUR
MOLEM48860052
Bis(4-hydroxyphenyl)sulfon
Bis(4-hydroxyphenyl)sulfon
Formel: C₁₂H₁₀O₄S
Molecular Weight: 250,28 g/mol
MDL: MFCD00002350
CAS-Nummer: 80-09-1
EINECS: 201-250-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR