4-Acetylbenzonitril
Lieferant: Molekula
Synonyme:
4'-Cyanacetophenon, Methyl-p-cyanphenylketon
M48915614
MOLEM48915614EA
0
EUR
MOLEM48915614
4-Acetylbenzonitril
4-Acetylbenzonitril
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol Siedepunkt: 110…112 °C (1 mmHg) Schmelzpunkt: 57…58 °C |
MDL:
MFCD00001825 CAS-Nummer: 1443-80-7 EINECS: 215-885-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...