Methyl-6-indolcarboxylat

Lieferant: Molekula

Synonyme: Methyl 1H-indole-6-carboxylate

M49212138
MOLEM49212138EA 0 EUR
MOLEM49212138
Methyl-6-indolcarboxylat
Methyl-6-indolcarboxylat
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
MDL: MFCD00211063
CAS-Nummer: 50820-65-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR