Fluorescein

Lieferant: Molekula

M49740067
MOLEM49740067EA 0 EUR
MOLEM49740067
Fluorescein
Fluorescein
Formel: C₂₀H₁₂O₅
Schmelzpunkt: 314…316 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005050
CAS-Nummer: 2321-07-5
EINECS: 219-031-8
Merck Index: 13,04185
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97b9d8d37f0106f4c8ef2d7759759d21bd43cc03&vid=25b78df95a04ab26c5ba8c96b95f031056926b13&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b0e97bec9c9f44dd58e1cd0ca79d7e94c3ba82d&vid=a29b350acb5fd71b32a30baa408af54494e42d59&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR