3-Chlor-5-fluoranilin

Lieferant: Molekula

M50661002
MOLEM50661002EA 0 EUR
MOLEM50661002
3-Chlor-5-fluoranilin
3-Chlor-5-fluoranilin
Formel: C₆H₅ClFN
MDL: MFCD03407962
CAS-Nummer: 4863-91-6
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR