Neocuproin
Lieferant: Molekula
Synonyme:
2,9-Dimethyl-1,10-phenanthrolin
M51096301
MOLEM51096301EA
0
EUR
MOLEM51096301
Neocuproin
Neocuproin
Formel:
C₁₄H₁₂N₂ Molecular Weight: 208,26 g/mol Schmelzpunkt: 161…163 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004973 CAS-Nummer: 484-11-7 EINECS: 207-601-9 Merck Index: 13,06477 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...