2-Naphthylboronsäure

Lieferant: Molekula
Achtung

Synonyme: 2-Naphthaleneboronic acid, Naphthalene-2-boronic acid, 2-Naphthyl boric acid

M51174107
MOLEM51174107EA 0 EUR
MOLEM51174107
2-Naphthylboronsäure
2-Naphthylboronsäure
Formel: C₁₀H₉BO₂
Molecular Weight: 171.99 g/mol
MDL: MFCD00236051
CAS-Nummer: 32316-92-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR