2-Mercaptobenzoxazol

Lieferant: Molekula

Synonyme: Benzo[d]oxazole-2-thiol

M51321929
MOLEM51321929EA 0 EUR
MOLEM51321929
2-Mercaptobenzoxazol
2-Mercaptobenzoxazole
Formel: C₇H₅NOS
Molecular Weight: 151,19 g/mol
MDL: MFCD00005769
CAS-Nummer: 2382-96-9
EINECS: 219-191-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR