6-Chloroxindol

Lieferant: Molekula

Synonyme: 6-Chloro-2-oxindole

M51694620
MOLEM51694620EA 0 EUR
MOLEM51694620
6-Chloroxindol
6-Chloroxindol
Formel: C₈H₆ClNO
Molecular Weight: 167,59 g/mol
MDL: MFCD00209962
CAS-Nummer: 56341-37-8
EINECS: 421-330-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR