D(+)-Prolin

Lieferant: Molekula

M51773021
MOLEM51773021EA 0 EUR
MOLEM51773021
D(+)-Prolin
D(+)-Prolin
Formel: C₅H₉NO₂
Schmelzpunkt: 223 °C
MDL: MFCD00064317
CAS-Nummer: 344-25-2
EINECS: 206-452-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR