2-Chlor-4-(trifluormethyl)benzonitril

Lieferant: Molekula

Synonyme: 2-Chlor-4-(trifluormethyl)benzoesäurenitril, 3-Chlor-4-cyanbenzotrifluorid

M52180013
MOLEM52180013EA 0 EUR
MOLEM52180013
2-Chlor-4-(trifluormethyl)benzonitril
2-Chlor-4-(trifluormethyl)benzonitril
Formel: C₈H₃ClF₃N
Molecular Weight: 205,57 g/mol
MDL: MFCD00084939
CAS-Nummer: 1813-33-8
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR