2-Amino-6-chlorpurin

Lieferant: Molekula

Synonyme: 6-Chlor-7H-purin-2-ylamin

M52216629
MOLEM52216629EA 0 EUR
MOLEM52216629
2-Amino-6-chlorpurin
2-Amino-6-chlorpurin
Formel: C₅H₄ClN₅
Molecular Weight: 169,57 g/mol
Schmelzpunkt: 300 °C
MDL: MFCD00075252
CAS-Nummer: 10310-21-1
EINECS: 233-686-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR