1-(Brommethyl)naphthalin

Lieferant: Molekula

M52360249
MOLEM52360249EA 0 EUR
MOLEM52360249
1-(Brommethyl)naphthalin
1-(Bromomethyl)naphthalene
Formel: C₁₁H₉Br
Molecular Weight: 221,10 g/mol
MDL: MFCD00010804
CAS-Nummer: 3163-27-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR