Anethol
Lieferant: Molekula
Synonyme:
cis-Anethol, 1-Methoxy-4-(prop-1-en-1-yl)benzene
M52441337
MOLEM52441337EA
560
EUR
MOLEM52441337
Anethol
Anethol
Formel:
C₁₀H₁₂O Molecular Weight: 148,2 g/mol Siedepunkt: 234,5 °C (1013 hPa) Schmelzpunkt: 21,3 °C Dichte: 1 g/cm³ (12 °C) |
MDL:
MFCD00009284 CAS-Nummer: 104-46-1 EINECS: 203-205-5 Merck Index: 13,00647 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...