Diosmin
Lieferant: Molekula
Synonyme:
5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one
M52585382
MOLEM52585382EA
0
EUR
MOLEM52585382
Diosmin
Diosmin
Formel:
C₂₈H₃₂O₁₅ |
MDL:
MFCD00009772 CAS-Nummer: 520-27-4 EINECS: 208-289-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...