2-Bromthiophenol
Lieferant: Molekula
Synonyme:
1-Brom-2-mercaptobenzol, 2-Brombenzol-1-thiol, o-Bromthiophenol, 2-Brombenzolthiol, 2-Bromphenylmercaptan
M52992637
MOLEM52992637EA
0
EUR
MOLEM52992637
2-Bromthiophenol
2-Bromthiophenol
Formel:
C₆H₅BrS Molecular Weight: 189.08 g/mol Siedepunkt: 128 Dichte: 1.604 g/cm³ (20 °C) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00004827 CAS-Nummer: 6320-02-1 EINECS: 228-665-4 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...