Nitroprussid-Natrium Dihydrat

Lieferant: Molekula
Gefahr

Synonyme: Dinatriumpentacyanonitrosylferrat(III) Dihydrat, Natriumnitrosopentacyanoferrat(III) Dihydrat, di-Natriumnitrosylpentacyanoferrat(III) Dihydrat, Natriumnitroprussiat Dihydrat, di-Natriumpentacyanonitrosylferrat(III) Dihydrat

M53304842
MOLEM53304842EA 0 EUR
MOLEM53304842
Nitroprussid-Natrium Dihydrat
Nitroprussid-Natrium Dihydrat
Formel: Na₂[Fe(CN)₅NO]·2H₂O
Molecular Weight: 297,95 g/mol
Dichte: 1,72 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149192
CAS-Nummer: 13755-38-9
EINECS: 238-373-9
UN: 1588
ADR: 6.1,III
Merck Index: 13,08722

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR