Dimethyl-5-nitroisophthalat
Lieferant: Molekula
Synonyme:
5-Nitro-1,3-benzoldicarbonsäuredimethylester, 5-Nitroisophthalsäuredimethylester
M54542162
MOLEM54542162EA
0
EUR
MOLEM54542162
Dimethyl-5-nitroisophthalat
Dimethyl-5-nitroisophthalat
Formel:
C₁₀H₉NO₆ Molecular Weight: 239,18 g/mol |
MDL:
MFCD00008429 CAS-Nummer: 13290-96-5 EINECS: 236-307-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...