3-Amino-2-methylbenzotrifluorid
Lieferant: Molekula
Synonyme:
2-Methyl-3-(trifluoromethyl)aniline
M54855298
MOLEM54855298EA
0
EUR
MOLEM54855298
3-Amino-2-methylbenzotrifluorid
3-Amino-2-methylbenzotrifluorid
Formel:
C₈H₈F₃N Molecular Weight: 175,16 g/mol |
MDL:
MFCD00153216 CAS-Nummer: 54396-44-0 EINECS: 259-145-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...