5-(Hydroxymethyl)resorcin

Lieferant: Molekula

Synonyme: (3,5-Dihydroxyphenyl)methanol, 3,5-Dihydroxybenzylalkohol

M55042890
MOLEM55042890EA 0 EUR
MOLEM55042890
5-(Hydroxymethyl)resorcin
5-(Hydroxymethyl)resorcin
Formel: C₇H₈O₃
Molecular Weight: 140,14 g/mol
MDL: MFCD00016867
CAS-Nummer: 29654-55-5
EINECS: 249-751-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR