5-Nitro-1H-indazol

Lieferant: Molekula

Synonyme: 5-NINA, 5-Nitro-1H-indazol

M55320728
MOLEM55320728EA 0 EUR
MOLEM55320728
5-Nitro-1H-indazol
5-Nitro-1H-indazole
Formel: C₇H₅N₃O₂
Molecular Weight: 163,14 g/mol
Schmelzpunkt: 210…212 °C
MDL: MFCD00005693
CAS-Nummer: 5401-94-5
EINECS: 226-451-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR