1,3-Diethinylbenzol

Lieferant: Molekula

M55367264
MOLEM55367264EA 0 EUR
MOLEM55367264
1,3-Diethinylbenzol
1,3-Diethynylbenzene
Formel: C₁₀H₆
MDL: MFCD00041686
CAS-Nummer: 1785-61-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR