4-Methyl-3-(trifluormethyl)benzoesäure

Lieferant: Molekula

M56718436
MOLEM56718436EA 0 EUR
MOLEM56718436
4-Methyl-3-(trifluormethyl)benzoesäure
4-Methyl-3-(trifluoromethyl)benzoic acid
Formel: C₉H₇F₃O₂
MDL: MFCD01631592
CAS-Nummer: 261952-01-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR