(Diacetoxyiod)benzol
Lieferant: Molekula
Synonyme:
Iodobenzene diacetate, (Diacetoxyiodo)benzene
M57233610
MOLEM57233610EA
0
EUR
MOLEM57233610
(Diacetoxyiod)benzol
(Diacetoxyiod)benzol
Formel:
C₁₀H₁₁IO₄ Molecular Weight: 322.1 g/mol |
MDL:
MFCD00008692 CAS-Nummer: 3240-34-4 EINECS: 221-808-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...