6-Bromindol

Lieferant: Molekula

Synonyme: 6-Bromo-1H-indole

M57350421
MOLEM57350421EA 0 EUR
MOLEM57350421
6-Bromindol
6-Bromindol
Formel: C₈H₆BrN
Lagertemperatur: Raumtemperatur
MDL: MFCD00238550
CAS-Nummer: 52415-29-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR