L(+)-Diisopropyltartrat
Lieferant: Molekula
Synonyme:
Diisopropyl (2R,3R)-2,3-dihydroxysuccinate, L-(+)-Tartaric acid diisopropyl ester
M57437082
MOLEM57437082EA
0
EUR
MOLEM57437082
L(+)-Diisopropyltartrat
L(+)-Diisopropyltartrat
Formel:
C₁₀H₁₈O₆ Molecular Weight: 234,25 g/mol |
MDL:
MFCD00064450 CAS-Nummer: 2217-15-4 EINECS: 218-709-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...