D-α-Tocopherol

Lieferant: Molekula

Synonyme: 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E Alkohol, (+)-α-Tocopherol, D(+)-α-Tocopherol, Vitamin E

M57476445
MOLEM57476445EA 0 EUR
MOLEM57476445
D-α-Tocopherol
D-α-Tocopherol
Formel: C₂₉H₅₀O₂
Molecular Weight: 430.71 g/mol
MDL: MFCD00072045
CAS-Nummer: 59-02-9
EINECS: 233-466-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR