5-Methoxy-1-tetralon

Lieferant: Molekula

Synonyme: 5-Methoxy-3,4-dihydronaphthalenone

M57513414
MOLEM57513414EA 0 EUR
MOLEM57513414
5-Methoxy-1-tetralon
5-Methoxy-1-tetralon
Formel: C₁₁H₁₂O₂
Molecular Weight: 176.22 g/mol
MDL: MFCD00001692
CAS-Nummer: 33892-75-0
EINECS: 251-723-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR