3,5-Dimethylisoxazol

Lieferant: Molekula

M57513896
MOLEM57513896EA 0 EUR
MOLEM57513896
3,5-Dimethylisoxazol
3,5-Dimethylisoxazole
Formel: C₅H₇NO
Molecular Weight: 97,12 g/mol
Siedepunkt: 142 °C (1013 hPa)
Dichte: 0,98 g/cm³ (20 °C)
MDL: MFCD00003156
CAS-Nummer: 300-87-8
EINECS: 206-100-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR