Bis(2-chlorethyl)ammoniumchlorid
Lieferant: Molekula
Synonyme:
Bis(2-chloroethyl)amine hydrochloride
M57679505
MOLEM57679505EA
0
EUR
MOLEM57679505
Bis(2-chlorethyl)ammoniumchlorid
Bis(2-chlorethyl)ammoniumchlorid
Formel:
C₄H₁₀Cl₃N Molecular Weight: 178,49 g/mol |
MDL:
MFCD00012515 CAS-Nummer: 821-48-7 EINECS: 212-479-5 UN: 2923 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...