D(-)-α-(4-Hydroxyphenyl)glycin
Lieferant: Molekula
Synonyme:
D(-)-4-Hydroxyphenylglycine, H-D-Phg(4-OH)-OH, (R)-(-)-2-(4-Hydroxyphenyl)glycine, (-)-4-Hydroxy-D-phenylglycine, D-(-)-p-Hydroxyphenylglycine
M57712989
MOLEM57712989EA
0
EUR
MOLEM57712989
D(-)-α-(4-Hydroxyphenyl)glycin
D(-)-α-(4-Hydroxyphenyl)glycin
Formel:
C₈H₉NO₃ Molecular Weight: 167,16 g/mol |
MDL:
MFCD00004262 CAS-Nummer: 22818-40-2 EINECS: 245-247-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...