D(-)-Cystein

Lieferant: Molekula

Synonyme: (S)-(-)-Cystein, H-D-Cys-OH, (S)-(-)-2-Amino-3-mercaptopropansäure

M57897158
MOLEM57897158EA 0 EUR
MOLEM57897158
D(-)-Cystein
D(-)-Cystein
Formel: C₃H₇NO₂S
Molecular Weight: 121,16 g/mol
MDL: MFCD00066461
CAS-Nummer: 921-01-7
EINECS: 213-062-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR