Sudan II

Lieferant: Molekula

M57901906
MOLEM57901906EA 0 EUR
MOLEM57901906
Sudan II
Sudan II
Formel: C₁₈H₁₆N₂O
MDL: MFCD00003896
CAS-Nummer: 3118-97-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR