D(-)-Glutamin

Lieferant: Molekula

Synonyme: (R)-(-)-Glutamin, H-D-Gln-OH, (R)-(-)-Glutaminsäure-5-amid, D(-)-Glutaminsäure-5-amid, (R)-(-)-2-Amino-1,5-pentandisäure-5-amid, D(-)-2-Amino-1,5-pentandisäure-5-amid

M58264131
MOLEM58264131EA 0 EUR
MOLEM58264131
D(-)-Glutamin
D(-)-Glutamin
Formel: C₅H₁₀N₂O₃
Molecular Weight: 146,15 g/mol
MDL: MFCD00065607
CAS-Nummer: 5959-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR