3-Brombiphenyl (PBB Nr. 2)
Lieferant: Molekula
Synonyme:
1-Brom-3-phenylbenzol
M58329326
MOLEM58329326EA
0
EUR
MOLEM58329326
3-Brombiphenyl (PBB Nr. 2)
3-Brombiphenyl (PBB Nr. 2)
Formel:
C₁₂H₉Br Molecular Weight: 233,11 g/mol Siedepunkt: 299 °C (1013 hPa) Schmelzpunkt: 9 °C Dichte: 1,41 g/cm³ (20 °C) |
MDL:
MFCD00000082 CAS-Nummer: 2113-57-7 EINECS: 218-304-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...