3,3'-Diaminobenzidin

Lieferant: Molekula

Synonyme: DAB, Biphenyl-3,3',4,4'-tetrayltetraamin

M58703130
MOLEM58703130EA 0 EUR
MOLEM58703130
3,3'-Diaminobenzidin
3,3'-Diaminobenzidin
Formel: C₁₂H₁₄N₄
Molecular Weight: 214,27 g/mol
MDL: MFCD00007725
CAS-Nummer: 91-95-2
EINECS: 202-110-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR