Tetrahydro-4H-pyran-4-on

Lieferant: Molekula

Synonyme: Tetrahydro-γ-pyran-4-on, Tetrahydro-2H-pyran-4-on, Tetrahydro-α-pyran-4-on

M58812912
MOLEM58812912EA 0 EUR
MOLEM58812912
Tetrahydro-4H-pyran-4-on
Tetrahydro-4H-pyran-4-on
Formel: C₅H₈O₂
Molecular Weight: 100,12 g/mol
Siedepunkt: 166 °C (1013 hPa)
Dichte: 1,08 g/cm³ (20 °C)
MDL: MFCD00006581
CAS-Nummer: 29943-42-8
EINECS: 249-967-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR