Phenyl-β-D-thioglucopyranosid

Lieferant: Molekula

Synonyme: (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(phenylthio)tetrahydro-2H-pyran-3,4,5-triol

M59128826
MOLEM59128826EA 0 EUR
MOLEM59128826
Phenyl-β-D-thioglucopyranosid
Phenyl-β-D-thioglucopyranosid
Formel: C₁₂H₁₆O₅S
Molecular Weight: 272,317 g/mol
MDL: MFCD11045365
CAS-Nummer: 2936-70-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR