(2R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamid
Lieferant: Molekula
Synonyme:
Dexpanthenol, (R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
M59191714
MOLEM59191714EA
0
EUR
MOLEM59191714
(2R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamid
D-Panthenol
Formel:
C₉H₁₉NO₄ Molecular Weight: 205.25 g/mol |
MDL:
MFCD00065006 CAS-Nummer: 81-13-0 EINECS: 201-327-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...